siRNA Details
| Gene Symbol: Gpr137b-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK170650.1 | - | 5 | 2008 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr137b-ps| Gene Symbol: Gpr137b-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK170650.1 | - | 5 | 2008 | ||