siRNA Details
Gene Symbol: 4930593A02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045167.1 | 7 | 516 | |||
GenBank mRNA | AK016388.1 | - | 7 | 517 | ||
AK017132.1 | - | 216 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930593A02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045167.1 | 7 | 516 | |||
GenBank mRNA | AK016388.1 | - | 7 | 517 | ||
AK017132.1 | - | 216 |