siRNA Details
| Gene Symbol: 9530036O11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK053224.1 | - | 761 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB535728.2 | - | 6 | 563 | Chr. 5 - 28564358 - 28933995 [+] on Build GRCm38 | |

9530036O11Rik
Genomic Map