siRNA Details
| Gene Symbol: 2310015A10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033514.1 | 3 | 2878 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2310015A10Rik| Gene Symbol: 2310015A10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033514.1 | 3 | 2878 | |||