siRNA Details
| Gene Symbol: 9530059O14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015610.1 | 1 | 247 | |||
| GenBank mRNA | AK048438.1 | - | 1 | 248 | ||
| AK079255.1 | - | 247 | ||||
| AK084074.1 | - | 1 | 247 | |||
| AK135014.1 | - | 251 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9530059O14Rik| Gene Symbol: 9530059O14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015610.1 | 1 | 247 | |||
| GenBank mRNA | AK048438.1 | - | 1 | 248 | ||
| AK079255.1 | - | 247 | ||||
| AK084074.1 | - | 1 | 247 | |||
| AK135014.1 | - | 251 | ||||