siRNA Details
| Gene Symbol: E030044B06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK086879.1 | - | 3 | 988 | ||
| BB530208.2 | - | 1 | 402 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E030044B06Rik| Gene Symbol: E030044B06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK086879.1 | - | 3 | 988 | ||
| BB530208.2 | - | 1 | 402 | |||