siRNA Details
Gene Symbol: Gm6225 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033457.1 | 2 | 997 | |||
GenBank mRNA | AK086784.1 | - | 2 | 734 | ||
AK163460.1 | - | 2 | 717 | |||
AK166419.1 | - | 2 | 723 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm6225 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033457.1 | 2 | 997 | |||
GenBank mRNA | AK086784.1 | - | 2 | 734 | ||
AK163460.1 | - | 2 | 717 | |||
AK166419.1 | - | 2 | 723 |