siRNA Details
| Gene Symbol: Gm16938 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045969.1 | 1 | 229 | |||
| NR_045970.1 | 1 | 229 | ||||
| NR_045971.1 | 1 | 229 | ||||
| NR_045972.1 | 1 | 229 | ||||
| GenBank mRNA | AK085722.1 | - | 230 | |||
| BC144932.1 | - | 1 | 182 | |||
| BC144933.1 | - | 1 | 182 | |||
| BC144936.1 | - | 1 | 182 | |||

Gm16938
Genomic Map