siRNA Details
| Gene Symbol: 4732463B04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK132490.1 | - | 3 | 2740 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4732463B04Rik| Gene Symbol: 4732463B04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK132490.1 | - | 3 | 2740 | ||