siRNA Details
| Gene Symbol: LOC73899 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014661.1 | - | 2 | 539 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714157.1 | - | 2 | 539 | Chr. 19 - 53460622 - 53462163 [+] on Build GRCm38 | |

LOC73899
Genomic Map