siRNA Details
| Gene Symbol: Lppos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK039760.1 | - | 995 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lppos| Gene Symbol: Lppos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK039760.1 | - | 995 | |||