siRNA Details
Gene Symbol: Celrr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038008.1 | 1 | 122 | |||
NR_110450.1 | 1 | 122 | ||||
GenBank mRNA | AK045543.1 | - | 55 | |||
AK138346.1 | - | 1 | 123 | |||
BE654424.1 | - | 1 | 33 |
