siRNA Details
Gene Symbol: Celrr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038008.1 | 5 | 1328 | |||
NR_110450.1 | 3 | 1067 | ||||
GenBank mRNA | AK138346.1 | - | 5 | 1329 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Celrr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038008.1 | 5 | 1328 | |||
NR_110450.1 | 3 | 1067 | ||||
GenBank mRNA | AK138346.1 | - | 5 | 1329 |