siRNA Details
| Gene Symbol: 4930455B14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045968.1 | 3 | 323 | |||
| GenBank mRNA | AK006409.1 | - | 3 | 324 | ||
| AK015458.1 | - | 3 | 268 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930455B14Rik| Gene Symbol: 4930455B14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045968.1 | 3 | 323 | |||
| GenBank mRNA | AK006409.1 | - | 3 | 324 | ||
| AK015458.1 | - | 3 | 268 | |||