siRNA Details
| Gene Symbol: 1700039M10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006644.1 | - | 2 | 402 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706633.1 | - | 2 | 402 | Chr. 14 - 57571624 - 57572368 [+] on Build GRCm38 | |

1700039M10Rik
Genomic Map