siRNA Details
| Gene Symbol: A330074K22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110496.1 | 3 | 1942 | |||
| GenBank mRNA | AK039624.1 | - | 3 | 1943 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A330074K22Rik| Gene Symbol: A330074K22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110496.1 | 3 | 1942 | |||
| GenBank mRNA | AK039624.1 | - | 3 | 1943 | ||