siRNA Details
| Gene Symbol: A330074K22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110496.1 | 2 | 404 | |||
| GenBank mRNA | AK039624.1 | - | 2 | 405 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CX214590.1 | - | 2 | 245 | Chr. 8 - 120206281 - 120228071 [-] on Build GRCm38 | |

A330074K22Rik
Genomic Map