siRNA Details
Gene Symbol: A330074K22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110496.1 | 3 | 2182 | |||
GenBank mRNA | AK039624.1 | - | 3 | 2183 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: A330074K22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110496.1 | 3 | 2182 | |||
GenBank mRNA | AK039624.1 | - | 3 | 2183 |