siRNA Details
| Gene Symbol: Ptprv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF300701.1 | - | 33 | 4993 | ||
| AK156112.1 | - | 34 | 5519 | |||
| U36488.1 | - | 30 | 4678 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptprv| Gene Symbol: Ptprv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF300701.1 | - | 33 | 4993 | ||
| AK156112.1 | - | 34 | 5519 | |||
| U36488.1 | - | 30 | 4678 | |||