siRNA Details
| Gene Symbol: Gm5914 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK080383.1 | - | 2 | 302 | ||
| AK086852.1 | - | 2 | 342 | |||
| BB529891.2 | - | 2 | 353 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5914| Gene Symbol: Gm5914 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK080383.1 | - | 2 | 302 | ||
| AK086852.1 | - | 2 | 342 | |||
| BB529891.2 | - | 2 | 353 | |||