siRNA Details
| Gene Symbol: 1700029N11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045489.1 | 4 | 546 | |||
| GenBank mRNA | AK006512.1 | - | 4 | 546 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700029N11Rik| Gene Symbol: 1700029N11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045489.1 | 4 | 546 | |||
| GenBank mRNA | AK006512.1 | - | 4 | 546 | ||