siRNA Details
| Gene Symbol: 1700095B10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040675.1 | 4 | 922 | |||
| NR_040676.1 | 4 | 825 | ||||
| GenBank mRNA | AK006812.1 | - | 4 | 905 | ||
| AK007075.1 | - | 4 | 828 | |||
| AK015608.1 | - | 4 | 826 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700095B10Rik| Gene Symbol: 1700095B10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040675.1 | 4 | 922 | |||
| NR_040676.1 | 4 | 825 | ||||
| GenBank mRNA | AK006812.1 | - | 4 | 905 | ||
| AK007075.1 | - | 4 | 828 | |||
| AK015608.1 | - | 4 | 826 | |||