siRNA Details
| Gene Symbol: Gm4258 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BE332980.1 | - | 36 | |||
| Gene Symbol: A630083H20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK040011.1 | - | 1 | 51 | ||

Gm4258
Genomic Map