siRNA Details
| Gene Symbol: 1190002F15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037955.1 | 2 | 213 | |||
| NR_037956.1 | 2 | 238 | ||||
| GenBank mRNA | AI121041.1 | - | 2 | 174 | ||
| AK004447.1 | - | 2 | 214 | |||
| AK012387.1 | - | 2 | 152 | |||
| AK138072.1 | - | 3404 | ||||

1190002F15Rik
Genomic Map