siRNA Details
| Gene Symbol: 7630403G23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040744.1 | 3 | 927 | |||
| GenBank mRNA | AK020190.1 | - | 3 | 929 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

7630403G23Rik| Gene Symbol: 7630403G23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040744.1 | 3 | 927 | |||
| GenBank mRNA | AK020190.1 | - | 3 | 929 | ||