siRNA Details
Gene Symbol: 1110028F18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045470.1 | 2 | 304 | |||
GenBank mRNA | AK003944.1 | - | 2 | 305 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1110028F18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045470.1 | 2 | 304 | |||
GenBank mRNA | AK003944.1 | - | 2 | 305 |