siRNA Details
| Gene Symbol: 1110028F18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045470.1 | 1 | 134 | |||
| GenBank mRNA | AK003944.1 | - | 1 | 135 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1110028F18Rik| Gene Symbol: 1110028F18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045470.1 | 1 | 134 | |||
| GenBank mRNA | AK003944.1 | - | 1 | 135 | ||