siRNA Details
| Gene Symbol: 0610031O16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045760.1 | 2 | 243 | |||
| GenBank mRNA | AK002733.1 | - | 2 | 243 | ||
| BC044770.1 | - | 501 | ||||
| BC046436.1 | - | 501 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

0610031O16Rik| Gene Symbol: 0610031O16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045760.1 | 2 | 243 | |||
| GenBank mRNA | AK002733.1 | - | 2 | 243 | ||
| BC044770.1 | - | 501 | ||||
| BC046436.1 | - | 501 | ||||