siRNA Details
| Gene Symbol: 1700047G03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040447.1 | 3 | 606 | |||
| GenBank mRNA | AK006707.1 | - | 3 | 607 | ||
| BC027576.1 | - | 751 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU939168.1 | - | 3 | 574 | Chr. 15 - 11967098 - 11969991 [+] on Build GRCm38 | |

1700047G03Rik
Genomic Map