siRNA Details
| Gene Symbol: 1700047G03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040447.1 | 2 | 152 | |||
| GenBank mRNA | AK006707.1 | - | 2 | 153 | ||
| BC027576.1 | - | 297 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700047G03Rik| Gene Symbol: 1700047G03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040447.1 | 2 | 152 | |||
| GenBank mRNA | AK006707.1 | - | 2 | 153 | ||
| BC027576.1 | - | 297 | ||||