siRNA Details
| Gene Symbol: 5730419F03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017575.1 | - | 1115 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5730419F03Rik| Gene Symbol: 5730419F03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017575.1 | - | 1115 | |||