siRNA Details
| Gene Symbol: 2410007B07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040539.1 | 3 | 893 | |||
| GenBank mRNA | AK010436.1 | - | 3 | 893 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV454330.1 | - | 1 | 342 | Chr. 3 - 75647446 - 75647920 [-] on Build GRCm38 | |

2410007B07Rik
Genomic Map