siRNA Details
| Gene Symbol: 2410007B07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040539.1 | 3 | 721 | |||
| GenBank mRNA | AK010436.1 | - | 3 | 721 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2410007B07Rik| Gene Symbol: 2410007B07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040539.1 | 3 | 721 | |||
| GenBank mRNA | AK010436.1 | - | 3 | 721 | ||