siRNA Details
| Gene Symbol: 2410007B07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040539.1 | 3 | 556 | |||
| GenBank mRNA | AK010436.1 | - | 3 | 556 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY709869.1 | - | 3 | 554 | Chr. 3 - 75647647 - 75655731 [-] on Build GRCm38 | |

2410007B07Rik
Genomic Map