siRNA Details
| Gene Symbol: 1700110C19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045461.1 | 1 | 178 | |||
| GenBank mRNA | AK007157.1 | - | 1 | 179 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700110C19Rik| Gene Symbol: 1700110C19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045461.1 | 1 | 178 | |||
| GenBank mRNA | AK007157.1 | - | 1 | 179 | ||