siRNA Details
| Gene Symbol: 4930533D04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015953.1 | - | 2 | 570 | ||
| AK131685.1 | - | 2 | 594 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV039747.3 | - | 802 | Chr. - - [-] on Build | ||

4930533D04Rik
Genomic Map