siRNA Details
| Gene Symbol: 1700096J18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027883.1 | 1 | 88 | |||
| GenBank mRNA | AK006164.1 | - | 1 | 89 | ||
| AK007092.1 | - | 1 | 75 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700096J18Rik| Gene Symbol: 1700096J18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027883.1 | 1 | 88 | |||
| GenBank mRNA | AK006164.1 | - | 1 | 89 | ||
| AK007092.1 | - | 1 | 75 | |||