siRNA Details
| Gene Symbol: 1700084F23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045965.1 | 1 | 92 | |||
| GenBank mRNA | AK005629.1 | - | 1 | 67 | ||
| AK006991.1 | - | 1 | 93 | |||
| AK016373.1 | - | 1 | 64 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700084F23Rik| Gene Symbol: 1700084F23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045965.1 | 1 | 92 | |||
| GenBank mRNA | AK005629.1 | - | 1 | 67 | ||
| AK006991.1 | - | 1 | 93 | |||
| AK016373.1 | - | 1 | 64 | |||