siRNA Details
| Gene Symbol: 1700031L13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006457.1 | - | 2 | 726 | ||
| AK006583.1 | - | 2 | 725 | |||
| AV044120.2 | - | 1 | 221 | |||
| AV207487.1 | - | 1 | 203 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700031L13Rik| Gene Symbol: 1700031L13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006457.1 | - | 2 | 726 | ||
| AK006583.1 | - | 2 | 725 | |||
| AV044120.2 | - | 1 | 221 | |||
| AV207487.1 | - | 1 | 203 | |||