siRNA Details
| Gene Symbol: 1700030F04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045731.1 | 3 | 266 | |||
| GenBank mRNA | AK006540.1 | - | 3 | 267 | ||
| BY706558.1 | - | 3 | 267 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700030F04Rik| Gene Symbol: 1700030F04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045731.1 | 3 | 266 | |||
| GenBank mRNA | AK006540.1 | - | 3 | 267 | ||
| BY706558.1 | - | 3 | 267 | |||