siRNA Details
| Gene Symbol: 1700029J03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040494.1 | 3 | 559 | |||
| GenBank mRNA | AK006500.1 | - | 3 | 560 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706535.1 | - | 3 | 560 | Chr. 16 - 93396918 - 93458872 [-] on Build GRCm38 | |

1700029J03Rik
Genomic Map