siRNA Details
Gene Symbol: 1700022E09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040568.1 | 2 | 139 | |||
GenBank mRNA | AK006237.1 | - | 2 | 140 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706321.1 | - | 2 | 140 | Chr. 16 - 59466870 - 59469238 [-] on Build GRCm38 |
