siRNA Details
| Gene Symbol: 1700036G14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040542.1 | 3 | 437 | |||
| GenBank mRNA | AK006614.1 | - | 3 | 438 | ||
| BC048678.1 | - | 3 | 418 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700036G14Rik| Gene Symbol: 1700036G14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040542.1 | 3 | 437 | |||
| GenBank mRNA | AK006614.1 | - | 3 | 438 | ||
| BC048678.1 | - | 3 | 418 | |||