siRNA Details
| Gene Symbol: Gm20324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045068.1 | 4 | 1076 | |||
| GenBank mRNA | AK080278.1 | - | 4 | 1078 | ||
| Gene Symbol: Tcap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC027631.1 | - | 883 | 1314541 | ||
| BC032992.1 | - | 883 | ||||

Gm20324
Genomic Map