siRNA Details
| Gene Symbol: 9930111H07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108086.1 | 4 | 668 | |||
| GenBank mRNA | AK020686.1 | - | 4 | 669 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9930111H07Rik| Gene Symbol: 9930111H07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108086.1 | 4 | 668 | |||
| GenBank mRNA | AK020686.1 | - | 4 | 669 | ||