siRNA Details
| Gene Symbol: 2610100L16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033490.1 | 3 | 1340 | |||
| GenBank mRNA | AK163433.1 | - | 3 | 1213 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2610100L16Rik| Gene Symbol: 2610100L16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033490.1 | 3 | 1340 | |||
| GenBank mRNA | AK163433.1 | - | 3 | 1213 | ||