siRNA Details
| Gene Symbol: 1700031P21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045910.1 | 2 | 175 | |||
| GenBank mRNA | AK006587.1 | - | 2 | 176 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700031P21Rik| Gene Symbol: 1700031P21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045910.1 | 2 | 175 | |||
| GenBank mRNA | AK006587.1 | - | 2 | 176 | ||