siRNA Details
| Gene Symbol: 1700031P21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045910.1 | 3 | 284 | |||
| GenBank mRNA | AK006587.1 | - | 3 | 285 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706590.1 | - | 3 | 280 | Chr. 12 - 52599984 - 52602318 [-] on Build GRCm38 | |

1700031P21Rik
Genomic Map