siRNA Details
| Gene Symbol: 1700003I22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005643.1 | - | 2 | 318 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700003I22Rik| Gene Symbol: 1700003I22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005643.1 | - | 2 | 318 | ||