siRNA Details
| Gene Symbol: Gm28505 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK080902.1 | - | 4 | 654 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB326078.2 | - | 4 | 673 | Chr. 17 - 11082254 - 11089460 [-] on Build GRCm38 | |

Gm28505
Genomic Map